2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082542-1G 082542-10G
102837-564EA 127.69 USD
102837-564 102837-566
2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid ≥97%
2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂O₅
MDL Number: MFCD02677730
CAS Number: 117621-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR