4,5-Dibromo-2-thenoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4,5-Dibromo-2-thiophenecarboxylic acid, 4,5-Dibromothiophene-2-carboxylic acid

082515-1G 082515-10G
102837-504EA 72.31 USD
102837-504 102837-506
4,5-Dibromo-2-thenoic acid ≥97%
4,5-Dibromo-2-thenoic acid

Hazard: Irritant

Formula: C₅H₂Br₂O₂S
Melting Pt: 227-228°C
MDL Number: MFCD00039682
CAS Number: 6324-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR