2-Hydroxy-6-methylisonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-6-methylpyridine-4-carboxylic acid

082478-1G 082478-10G
102837-420EA 69.23 USD
102837-420 102837-422
2-Hydroxy-6-methylisonicotinic acid ≥97%
2-Hydroxy-6-methylisonicotinic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.15 g/mol
MDL Number: MFCD02682019
CAS Number: 86454-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR