3-Pyridin-2-ylacrylic acid ≥97%

Supplier: Matrix Scientific

082468-1G 082468-10G
102837-398EA 40 USD
102837-398 102837-400
3-Pyridin-2-ylacrylic acid ≥97%
3-Pyridin-2-ylacrylic acid

Hazard: Irritant

Formula: C₈H₇NO₂
MW: 149.15 g/mol
MDL Number: MFCD00179422
CAS Number: 7340-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR