2-(Ethylthio)nicotinic acid ≥97%
Supplier: Matrix Scientific
082454-1G
082454-10G
102837-368EA
72.31
USD
102837-368
102837-370
2-(Ethylthio)nicotinic acid ≥97%
2-(Ethylthio)nicotinic acid
Hazard: Irritant
Formula:
C₈H₉NO₂S |
MDL Number:
MFCD00051680 CAS Number: 27868-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...