3-(3-Pyridyl)propionic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Pyridinepropionic Acid, 3-Pyridinepropionic acid

082384-1G 082384-10G
102837-226EA 72.31 USD
102837-226 102837-228
3-(3-Pyridyl)propionic acid ≥97%
3-(3-Pyridyl)propionic acid

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00274197
CAS Number: 3724-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR