3-(6-Methoxy-3-nitropyridin-2-yl)-2-methylpropanoic acid ≥97%

Supplier: Matrix Scientific

082370-1G 082370-10G
102837-196EA 298.46 USD
102837-196 102837-198
3-(6-Methoxy-3-nitropyridin-2-yl)-2-methylpropanoic acid ≥97%
3-(6-Methoxy-3-nitropyridin-2-yl)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₅
MW: 240.22 g/mol
MDL Number: MFCD12827862
CAS Number: 175205-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR