Isonicotinic acid-N-oxide ≥97%

Supplier: Matrix Scientific

082367-1G 082367-10G 082367-50G
102837-186EA 44.62 USD
102837-186 102837-188 102837-190
Isonicotinic acid-N-oxide ≥97%
Isonicotinic acid N-oxide

Hazard: Irritant

Formula: C₆H₅NO₃
MDL Number: MFCD00006209
CAS Number: 13602-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR