2-(4-Chlorophenylthio)nicotinic acid ≥97%

Supplier: Matrix Scientific

082365-1G 082365-10G
102837-180EA 86.15 USD
102837-180 102837-182
2-(4-Chlorophenylthio)nicotinic acid ≥97%
2-(4-Chlorophenylthio)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₈ClNO₂S
MDL Number: MFCD00052117
CAS Number: 955-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR