2-(Propylthio)nicotinic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(n-Propylthio)nicotinic acid
082363-1G
082363-10G
102837-174EA
101.54
USD
102837-174
102837-176
2-(Propylthio)nicotinic acid ≥97%
2-(Propylthio)nicotinic acid
Hazard: Irritant
Formula:
C₉H₁₁NO₂S |
MDL Number:
MFCD00051682 CAS Number: 175135-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...