2-Ethoxy-5-iodonicotinic acid ≥97%

Supplier: Matrix Scientific

082357-1G 082357-10G
102837-160EA 283.08 USD
102837-160 102837-162
2-Ethoxy-5-iodonicotinic acid ≥97%
2-Ethoxy-5-iodonicotinic acid

Hazard: Irritant

Formula: C₈H₈INO₃
MDL Number: MFCD09836165
CAS Number: 335078-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR