Picloram ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3,5,6-trichloropicolinic Acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic Acid

082356-1G 082356-10G
102837-156EA 101.54 USD
102837-156 102837-158
Picloram ≥97%
Picloram

TCA listed: T
Hazard: Irritant

Formula: C₆H₃Cl₃N₂O₂
MW: 241.45 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00012101
CAS Number: 1918-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR