4-Methylnicotinic acid hydrochloride ≥97%

Supplier: Matrix Scientific

082284-1G 082284-10G
102837-010EA 200 USD
102837-010 102837-012
4-Methylnicotinic acid hydrochloride ≥97%
4-Methylnicotinic acid hydrochloride

Hazard: Irritant

Formula: C₇H₈ClNO₂
MDL Number: MFCD00832700
CAS Number: 94015-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR