6-Phenoxynicotinic acid ≥97%

Supplier: Matrix Scientific

082283-1G 082283-10G
102837-006EA 383.08 USD
102837-006 102837-008
6-Phenoxynicotinic acid ≥97%
6-Phenoxynicotinic acid

Hazard: Irritant

Formula: C₁₂H₉NO₃
MDL Number: MFCD02682068
CAS Number: 51362-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR