3-Hydroxy-2,4,6-triiodobenzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: HTBA

082281-1G 082281-10G 082281-50G
102836-998EA 184.62 USD
102836-998 102837-000 102837-002
3-Hydroxy-2,4,6-triiodobenzoic acid ≥97%
3-Hydroxy-2,4,6-triiodobenzoic acid

Hazard: Irritant

Formula: C₇H₃I₃O₃
MW: 515.81 g/mol
MDL Number: MFCD00020305
CAS Number: 53279-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR