4,4'-Stilbenedicarboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4,4-Stilbenedicarboxylic acid

082277-1G 082277-10G 082277-50G
102836-986EA 101.54 USD
102836-986 102836-988 102836-990
4,4'-Stilbenedicarboxylic acid ≥98%
4,4'-Stilbenedicarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
MDL Number: MFCD00013994
CAS Number: 100-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR