(2E)-3-(2,4-Dihydroxyphenyl)acrylic acid ≥97%

Supplier: Matrix Scientific

082265-1G 082265-10G
102836-958EA 123.08 USD
102836-958 102836-960
(2E)-3-(2,4-Dihydroxyphenyl)acrylic acid ≥97%
(2E)-3-(2,4-Dihydroxyphenyl)acrylic acid

Hazard: Irritant

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00463787
CAS Number: 614-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR