4-Amino-3-bromobenzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-bromobenzoic Acid

082264-1G 082264-10G
102836-954EA 226.15 USD
102836-954 102836-956
4-Amino-3-bromobenzoic acid ≥97%
4-Amino-3-bromobenzoic acid

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
MDL Number: MFCD03407439
CAS Number: 6311-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR