(4-Nitrophenyl)acetic acid ≥90%

Supplier: Matrix Scientific

082258-10G 082258-50G
102836-940EA 52.31 USD
102836-940 102836-942
(4-Nitrophenyl)acetic acid ≥90%
(4-Nitrophenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₄(NO₂))CH₂COOH
MDL Number: MFCD00007383
CAS Number: 104-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR