3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid ≥97%
Supplier: Matrix Scientific
082252-1G
082252-10G
102836-924EA
56.92
USD
102836-924
102836-926
3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid ≥97%
3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid
Hazard: Irritant
Formula:
C₉H₈ClNO₆ |
MDL Number:
MFCD00052955 CAS Number: 175135-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...