3,5-Diiodosalicylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-3,5-diiodobenzoic acid

082251-10G 082251-50G
102836-920EA 90.77 USD
102836-920 102836-922
3,5-Diiodosalicylic acid ≥97%
3,5-Diiodosalicylic acid

Hazard: Irritant

Formula: C₇H₄I₂O₃
MW: 389.92 g/mol
MDL Number: MFCD00002444
CAS Number: 133-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR