2-Methyl-3-(trifluoromethyl)benzoic acid ≥97%
Supplier: Matrix Scientific
082247-1G
082247-10G
102836-910EA
127.69
USD
102836-910
102836-912
2-Methyl-3-(trifluoromethyl)benzoic acid ≥97%
2-Methyl-3-(trifluoromethyl)benzoic acid
Hazard: Irritant
Formula:
C₉H₇F₃O₂ MW: 204.15 g/mol |
MDL Number:
MFCD01631588 CAS Number: 62089-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...