5-(Chlorosulfonyl)-2-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific

082235-1G 082235-10G
102836-880EA 156.92 USD
102836-880 102836-882
5-(Chlorosulfonyl)-2-fluorobenzoic acid ≥97%
5-(Chlorosulfonyl)-2-fluorobenzoic acid

Hazard: Irritant

Formula: C₇H₄ClFO₄S
MW: 238.613 g/mol
MDL Number: MFCD03030335
CAS Number: 37098-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR