2-((Difluoromethyl)thio)benzoic acid ≥97%

Supplier: Matrix Scientific

082229-1G 082229-10G
102836-866EA 101.54 USD
102836-866 102836-868
2-((Difluoromethyl)thio)benzoic acid ≥97%
2-((Difluoromethyl)thio)benzoic acid

Hazard: Irritant

Formula: C₈H₆F₂O₂S
MW: 204.2 g/mol
CAS Number: 79676-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR