3-Methyl-2-phenylpentanoic acid ≥97%

Supplier: Matrix Scientific

082212-1G 082212-10G
102836-824EA 184.62 USD
102836-824 102836-826
3-Methyl-2-phenylpentanoic acid ≥97%
3-Methyl-2-phenylpentanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
MDL Number: MFCD00004254
CAS Number: 7782-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR