2-(tert-Butoxycarbonyl)benzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Phthalic acid mono-tert-butyl ester

082157-1G 082157-10G
102836-704EA 76.92 USD
102836-704 102836-706
2-(tert-Butoxycarbonyl)benzoic acid ≥97%
2-(tert-Butoxycarbonyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 84…88 °C
MDL Number: MFCD00043711
CAS Number: 33693-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR