3-Cyclopentyloxy-4-methoxybenzoic acid ≥97%

Supplier: Matrix Scientific

082148-1G 082148-10G
102836-678EA 213.85 USD
102836-678 102836-680
3-Cyclopentyloxy-4-methoxybenzoic acid ≥97%
3-Cyclopentyloxy-4-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
CAS Number: 144036-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR