3,4-Dimethoxybenzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Veratric acid

082139-1G 082139-10G 082139-50G
102836-656EA 37.92 USD
102836-656 102836-658 102836-660
3,4-Dimethoxybenzoic acid ≥97%
3,4-Dimethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 303
MDL Number: MFCD00002500
CAS Number: 93-07-2
Merck Index: 12,10086

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR