3-(1H-Imidazol-1-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082127-1G 082127-10G
102836-630EA 298.46 USD
102836-630 102836-632
3-(1H-Imidazol-1-yl)benzoic acid ≥97%
3-(1H-Imidazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD06659077
CAS Number: 108035-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR