4-Pentylbenzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amylbenzoic acid, 4-n-Pentylbenzoic acid, 4-Pentylbenzoic acid

082115-1G 082115-10G 082115-50G
102836-592EA 44.62 USD
102836-592 102836-594 102836-596
4-Pentylbenzoic acid ≥97%
4-Pentylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
MDL Number: MFCD00002572
CAS Number: 26311-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR