3-Nitrosalicylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Nitrosalicylic Acid, 2-Hydroxy-3-nitrobenzoic acid
082101-1G
082101-10G
082101-50G
102836-560EA
46.15
USD
102836-560
102836-562
102836-564
3-Nitrosalicylic acid ≥97%
3-Nitrosalicylic acid
Hazard: Irritant
Formula:
C₇H₅NO₅ MW: 183.1183 g/mol |
MDL Number:
MFCD00024240 CAS Number: 85-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...