2-(Acetyloxy)-3-methylbenzoic acid ≥97%

Supplier: Matrix Scientific

082100-1G 082100-10G
102836-556EA 184.62 USD
102836-556 102836-558
2-(Acetyloxy)-3-methylbenzoic acid ≥97%
2-(Acetyloxy)-3-methylbenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD00085683
CAS Number: 4386-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR