3-Phenoxybenzoic acid ≥97%

Supplier: Matrix Scientific

082099-1G 082099-10G
102836-552EA 72.31 USD
102836-552 102836-554
3-Phenoxybenzoic acid ≥97%
3-Phenoxybenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
MDL Number: MFCD00002498
CAS Number: 3739-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR