3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid ≥97%

Supplier: Matrix Scientific

082098-1G 082098-10G
102836-548EA 383.08 USD
102836-548 102836-550
3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid ≥97%
3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₉H₈Br₂O₃
MDL Number: MFCD00816760
CAS Number: 13811-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR