3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid ≥97%

Supplier: Matrix Scientific

082095-1G 082095-10G
102836-538EA 161.54 USD
102836-538 102836-540
3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid ≥97%
3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₄
MDL Number: MFCD00173707
CAS Number: 175278-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR