4-(4-Octylphenyl)-4-oxobutanoic acid ≥97%

Supplier: Matrix Scientific

082094-1G 082094-10G
102836-534EA 298.46 USD
102836-534 102836-536
4-(4-Octylphenyl)-4-oxobutanoic acid ≥97%
4-(4-Octylphenyl)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₈H₂₆O₃
MDL Number: MFCD00173760
CAS Number: 64779-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR