N-(4-Bromophenyl)maleamic acid ≥95%

Supplier: Matrix Scientific

082085-1G 082085-10G
102836-514EA 353.85 USD
102836-514 102836-516
N-(4-Bromophenyl)maleamic acid ≥95%
N-(4-Bromophenyl)maleamic acid

Hazard: Irritant

Formula: C₁₀H₈BrNO₃
MDL Number: MFCD00134952
CAS Number: 36847-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR