[4-(Dimethylamino)phenyl]acetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Dimethylamino)phenylacetic acid

082077-1G 082077-10G
102836-494EA 164.62 USD
102836-494 102836-496
[4-(Dimethylamino)phenyl]acetic acid ≥97%
[4-(Dimethylamino)phenyl]acetic acid

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00075591
CAS Number: 17078-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR