2-(Acetylamino)-5-nitrobenzoic acid ≥97%

Supplier: Matrix Scientific

082066-1G 082066-10G
102836-468EA 127.69 USD
102836-468 102836-470
2-(Acetylamino)-5-nitrobenzoic acid ≥97%
2-(Acetylamino)-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₉H₈N₂O₅
MDL Number: MFCD00204175
CAS Number: 3558-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR