4-(1H-Imidazol-1-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(1-Imidazolyl)benzoic acid

082065-1G 082065-10G
102836-464EA 123.08 USD
102836-464 102836-466
4-(1H-Imidazol-1-yl)benzoic acid ≥97%
4-(1H-Imidazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00586645
CAS Number: 17616-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR