5-Nitrosalicylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-5-nitrobenzoic acid

082059-10G 082059-50G
102836-442EA 78.46 USD
102836-442 102836-444
5-Nitrosalicylic acid ≥97%
5-Nitrosalicylic acid

Hazard: Irritant

Formula: C₇H₅NO₅
MW: 183.12 g/mol
MDL Number: MFCD00007338
CAS Number: 96-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR