5-Acetylsalicylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Acetyl-2-hydroxybenzoic acid, 5-Acetylsalicylic Acid
082035-1G
082035-10G
082035-50G
102836-386EA
72.31
USD
102836-386
102836-388
102836-390
5-Acetylsalicylic acid ≥97%
5-Acetylsalicylic acid
Hazard: Irritant
Formula:
C₉H₈O₄ MW: 180.15 g/mol |
MDL Number:
MFCD00013978 CAS Number: 13110-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...