5-Hydroxy-2-nitrobenzoic acid ≥97%
Supplier: Matrix Scientific
082028-1G
082028-10G
102836-362EA
113.85
USD
102836-362
102836-364
5-Hydroxy-2-nitrobenzoic acid ≥97%
5-Hydroxy-2-nitrobenzoic acid
Hazard: Irritant
Formula:
C₇H₅NO₅ MW: 183.12 g/mol |
MDL Number:
MFCD00017566 CAS Number: 610-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...