2-Fluoro-5-(trifluoromethoxy)phenylboronic acid ≥98%

Supplier: Matrix Scientific

081994-1G 081994-10G
102836-286EA 60 USD
102836-286 102836-288
2-Fluoro-5-(trifluoromethoxy)phenylboronic acid ≥98%
2-Fluoro-5-(trifluoromethoxy)phenylboronic acid

Hazard: Irritant

Formula: C₇H₅BF₄O₃
MDL Number: MFCD09027234
CAS Number: 881402-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR