2-Phenoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Phenoxybenzeneboronic acid

081991-1G 081991-10G
102836-278EA 53.85 USD
102836-278 102836-280
2-Phenoxyphenylboronic acid ≥97%
2-Phenoxyphenylboronic acid

Hazard: Irritant

Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
MDL Number: MFCD01001592
CAS Number: 108238-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR