Phenylboronic acid neopentyl glycol ester ≥97%
Supplier: Matrix Scientific
Synonyms:
5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane
081990-1G
081990-10G
102836-274EA
76.92
USD
102836-274
102836-276
Phenylboronic acid neopentyl glycol ester ≥97%
Phenylboronic acid neopentyl glycol ester
Hazard: Irritant
Formula:
C₁₁H₁₅BO₂ MW: 190.05 g/mol |
MDL Number:
MFCD05663851 CAS Number: 5123-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...