2-(4-Benzylpiperazin-1-yl)ethanamine ≥97%

Supplier: Matrix Scientific

081958-1G 081958-10G
102836-204EA 143.08 USD
102836-204 102836-206
2-(4-Benzylpiperazin-1-yl)ethanamine ≥97%
2-(4-Benzylpiperazin-1-yl)ethanamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD00831097
CAS Number: 4553-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR