4-(Aminomethyl)piperidine ≥97%

Supplier: Matrix Scientific

081956-1G 081956-10G 081956-50G
102836-196EA 72.31 USD
102836-196 102836-198 102836-200
4-(Aminomethyl)piperidine ≥97%
4-(Aminomethyl)piperidine

Hazard: Irritant

Formula: C₆H₁₄N₂
MDL Number: MFCD00006007
CAS Number: 7144-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR