Benzyl-3-(aminomethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

081955-1G 081955-10G
102836-192EA 327.69 USD
102836-192 102836-194
Benzyl-3-(aminomethyl)piperidine-1-carboxylate ≥95%
Benzyl-3-(aminomethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MW: 248.33 g/mol
MDL Number: MFCD03001691
CAS Number: 315717-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR