1-(2,3-Dihydro-1-benzofuran-5-yl)methanamine ≥97%

Supplier: Matrix Scientific

081951-1G 081951-10G
102836-182EA 156.92 USD
102836-182 102836-184
1-(2,3-Dihydro-1-benzofuran-5-yl)methanamine ≥97%
1-(2,3-Dihydro-1-benzofuran-5-yl)methanamine

Hazard: Irritant

Formula: C₉H₁₁NO
MDL Number: MFCD00728869
CAS Number: 55745-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR